Midostaurin ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide

BML-EI370-0001 BML-EI370-0005
89158-210EA 257.54 CAD
89158-210 89158-212
Midostaurin ≥98% (by TLC)
Midostaurin

Inhibitor of a variety of serine/threonine and tyrosine kinases, like protein kinase C (PKC), cyclic AMP-dependent protein kinase (PKA), S6 kinase, Akt (protein kinase B; PKB), epidermal growth factor receptor (EGFR) tyrosine kinase activity and others including KDR, VEGFR, PDGFR, c-kit and other receptor tyrosine kinases. Potently inhibits FLT-3 kinase including mutant forms found in acute myeloid leukemia in vitro and in vivo. Apoptosis inducer. Showed broad antiproliferative activity against various tumor cell lines. Selectively inhibits T lymphocyte production of TNF-α. Upregulates endothelial nitric oxide synthase (eNOS; NOS III). Abrogates tumor angiogenesis in vivo.


  • Flt inhibitor


White to off-white powder.


Soluble in DMSO (15mg/ml), MDC (10mg/ml), methanol (5mg/ml) or 100% ethanol (2.5mg/ml).

Formula: C₃₅H₃₀N₄O₄
MW: 570.64 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12828879
CAS Number: 120685-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0062b2ccc4f8508f4473e7c42dead7e614f39a28&vid=23cd6abc119b8a130f604ca06902cf82b3c87193&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR