STAT3 Inhibitor V, Stattic ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Stattic, Stat3 Inhibitor V, 6-Nitrobenzo[b]thiophene 1,1-dioxide, 6-​Nitro-​1,​1-​dioxide-​benzo[b]thiophene, 6-Nitro[b]thiophene-1,1-dioxide

BML-EI368-0010 BML-EI368-0050
89158-202EA 116.92 CAD
89158-202 89158-204
STAT3 Inhibitor V, Stattic ≥98% (by TLC)
STAT3 Inhibitor V, Stattic

Small molecule STAT3-specific inhibitor which blocks activation, dimerization and nuclear translocation via binding to the STAT3 SH2 domain. It is a very useful tool for exploring STAT3-mediated signaling pathways. It blocks LPS-mediated STAT3 tyrosine phosphorylation which leads to inhibition of LPS-mediated IL-1β and IL-6 production. In primary cultures of rat neural stem cells it blocks IL-15-induced differentiation.


  • STAT inhibitor


Pale yellow solid.


Soluble in DMSO (>25 mg/ml), 100% ethanol (10 mg/ml).

Formula: C₈H₅NO₄S
Storage Temperature: Ambient
MDL Number: MFCD00173799
CAS Number: 19983-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e6d515980289720d217e0e18de9826b1441370f&vid=945093cc2b2f191ed8c99492c1668b33c1633a78&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR