AAL-993 ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
AAL 993, 2-[(4-Pyridyl)methyl]amino-N-[3-(trifluoromethyl)phenyl]benzamide, 2-[(4-Pyridinylmethyl)amino]-N-[3-(trifluoromethyl)phenyl]benzamide
BML-EI367-0005
BML-EI367-0025
89158-198EA
325.48
CAD
89158-198
89158-200
AAL-993 ≥98% (by HPLC)
AAL-993
Highly potent inhibitor of VEGFR-1 (IC50=130nM), VEGFR-2 (IC50=23nM) and VEGFR-3 (IC50=18nM). At higher concentrations it inhibits PDGFR-β (640nM), c-Kit (236nM) and CSF-1R (380nM).
- VEGFR inhibitor
Pale yellow solid.
Soluble in DMSO (>25 mg/ml) or ethanol (15 mg/ml).
Formula:
C₂₀H₁₆F₃N₃O MW: 371.36 g/mol Storage Temperature: Ambient |
CAS Number:
269390-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...