GW-441756 ≥98%
Supplier: Enzo Life Sciences
BML-EI364-0010
BML-EI364-0050
89158-184EA
325.48
CAD
89158-184
89158-186
GW-441756 ≥98%
GW-441756
TrkA inhibitor
Potent and selective inhibitor of NGFR kinase (TrkA, IC50=2 nM). Blocks TrkA-mediated γH2AX production and apoptosis in TrkA-overexpressing cells.
Formula:
C₁₇H₁₃N₃O |
MDL Number:
MFCD17010281 CAS Number: 504433-23-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...