PD-146176 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI356-0010 BML-EI356-0050
89158-168EA 192.76 CAD
89158-168 89158-170
PD-146176 ≥98% (by HPLC)
PD-146176

A potent and selective 15-lipoxygenase inhibitor which lacks significant nonspecific antioxidant properties. It suppresses atherogenesis and limits atherosclerotic lesion development in the rabbit. It reduces oxidant stress-induced apoptosis in endothelial cells and inhibits proliferation in 15-LO overexpressing PC3 cells.


  • Lipoxygenase inhibitor


A potent and selective 15-lipoxygenase inhibitor which lacks significant nonspecific antioxidant properties. It suppresses atherogenesis and limits atherosclerotic lesion development in the rabbit. It reduces oxidant stress-induced apoptosis in endothelial cells and inhibits proliferation in 15-LO overexpressing PC3 cells.

Formula: C₁₅H₁₁NS
Storage Temperature: Ambient
MDL Number: MFCD05664738
CAS Number: 4079-26-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b434f9dc23eca19c05e4b9582c8503e4f8df9c77&vid=85b137cb48fcaa5022b1a53306dcfd8908009063&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR