Apigenin ≥97% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one

BML-EI345-0020 BML-EI345-0100
89158-136EA 173.8 CAD
89158-136 89158-138
Apigenin ≥97% (by TLC)
Apigenin

Antioxidant and chemopreventive


Apigenin is an antioxidant flavonoid. Has chemopreventive and antitumor  properties. Induces apoptosis. Inhibits the proliferation of malignant tumor cells by G2/M arrest. Is a MAP-Kinase (MAPK/ERK) inhibitor. Inhibits hypoxia-inducible factor-1 (HIF-1) and vascular endothelial growth factor (VEGF) expression. Induces gastric relaxation in mouse stomach, mainly by suppressing Ca2+ influx through voltage-dependent calcium channels. Apigenin is a selective inhibitor of CK2. In WEHI 231 cells inhibition of CK2 by apigenin decreased the rate of IκBα turnover and nuclear levels of NF-κB (20 µM). It blocks proliferation of Wnt-1 transfected cells and abrogates phosphorylation of β-catenin. In a rat model of glomerulonephritis, inhibition of CK2 by apigenin ameliorates renal dysfunction.

Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
MDL Number: MFCD00006831
CAS Number: 520-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dad25b9042b65d3156a74ecf2c1e17a39e757284&vid=8b5db294fc5cd22fe2317bee7af2ae30e28f479f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR