RO 48-8071 ≥95% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI342-0010 BML-EI342-0050
89158-124EA 162.74 CAD
89158-124 89158-126
RO 48-8071 ≥95% (by HPLC)
RO 48-8071

Inhibits cholesterol biosynthesis at the level of 2,3-oxidosqualene cyclase (OSC). Blocks OSC and cholesterol synthesis in HepG2 cells in the nanomolar range. A novel lipid lowering agent.


  • Oxido-squalene cyclase inhibitor


Pale yellow solid.


Soluble in water (25 mg/ml)

Formula: C₂₃H₂₇BrFNO₂.C₄H₄O₄
Storage Temperature: Ambient
MDL Number: MFCD05865242
CAS Number: 189197-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5cb77a4ad0172ef14421d20dbff445f65403837&vid=8feafbbb2a008a3b4c0c79664ccc375959114654&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR