Triciribine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI332-0001 BML-EI332-0005
89158-096EA 357.08 CAD
89158-096 89158-098
Triciribine ≥98% (by HPLC)
Triciribine

Akt inhibitor


Triciribine is a synthetic tricyclic nucleoside which acts as a specific inhibitor of the Akt signaling pathway. It selectively inhibits the phosphorylation and activation of Akt1, -2 and -3 but does not inhibit Akt kinase activity nor known upstream Akt activators such as PI 3-Kinase and PDK1. It inhibits cell growth and induces apoptosis preferentially in cells that express aberrant Akt1. In whole cells triciribine is phosphorylated by adenosine kinase which may be necessary for its activity.

Formula: C₁₃H₁₆N₆O₄
MDL Number: MFCD00932413
CAS Number: 35943-35-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e4405096c77b5d29bbac10bb4c50df8bf26dbc4&vid=cee53cd27b6569ae4041950adb3a307041c1b64c&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR