5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥99% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: AICAR, 5-Amino-1-(β-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine

BML-EI330-0050 BML-EI330-0250
89158-088EA 176.96 CAD
89158-088 89158-090
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥99% (by TLC)
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside

AMPK activator


An adenosine analog that is phosphorylated in whole cells to form 5-aminoimidazole-4-carboxamide-1-D-ribofuranosyl-5’-monophosphate (ZMP), which stimulates AMPK activity. AICAR acts by entering nucleoside pools, significantly increasing levels of adenosine during periods of ATP breakdown. Induces the phosphorylation of TORC2 and inhibits its entry into the nucleus. Mimics the effects of insulin on the expression of two gluconeogenic genes PEPCK and glucose-6-phosphatase. Inhibits PPARα coactivation and adipocyte differentiation. Substrate for the AICAR transformylase activity of ATIC. Inhibits the growth of prostate cancer cells by inhibiting fatty ascid synthesis. Supresses LPS-induced TNF production via the inhibition of Akt activation. Induces apoptosis in Jurkat cells via a non-AMPK pathway.

Formula: C₉H₁₄N₄O₅
MW: 258.23 g/mol
MDL Number: MFCD00869751
CAS Number: 2627-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4f853de4791186729bc93c216606098a189e09f&vid=e95f6f88a15e44802930e87b09138748f32505ce&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR