TOFA ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-EI321-0010 BML-EI321-0050
89158-066EA 153.26 CAD
89158-066 89158-068
TOFA ≥99% (by TLC)
TOFA

Acetyl-CoA inhibitor


Hypolipidemic agent. Potent, reversible and competitive inhibitor of acetyl-coenzyme A carboxylase (Ac-CoA carboxylase; ACC), a key enzyme involved in fatty acid biosynthesis. It blocks apoptosis induced by the fatty acid synthase inhibitor C75 (Prod. No. ALX-270-286) and stimulates neurite outgrowth and neuronal differentiation in rat pheochromocytoma cells.

Formula: C₁₉H₃₂O₄
MDL Number: MFCD01726059
CAS Number: 54857-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6e6fa16b9857340b82df751f5cb9017aa59389f&vid=1d4a03e744c1b07a8f6758c144c1e52543d02907&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR