TMP-153 ≥98%

Supplier: Enzo Life Sciences

BML-EI317-0020 BML-EI317-0100
89158-056EA 140.62 CAD
89158-056 89158-058
TMP-153 ≥98%
TMP-153

TMP-153 is a potent and selective inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT), IC50=5-10 nM. It inhibits cholesterol absorption and lowers plasma cholesterol in rats and hamsters. ACAT inhibitors improve plasma lipid profile and ameliorate proteinuria in nephrotic animals.


  • ACAT inhibitor


White solid.


Soluble in DMSO (>25mg/ml) or ethanol (10mg/ml with warming).

Formula: C₂₄H₁₈ClF₂N₃O
Storage Temperature: Ambient
CAS Number: 128831-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bc2a4b87e66c6b24f66dbc7635c5c83a972b6b6&vid=26bfa933b594b13d81d067396aa761e0d417fb71&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR