SU4312 ≥98%

Supplier: Enzo Life Sciences

BML-EI306-0005 BML-EI306-0025
89158-022EA 180.12 CAD
89158-022 89158-024
SU4312 ≥98%
SU4312

Potent and selective inhibitor of VEGFR-2 (Flk-1, KDR). It displays a 100-fold greater potency for the unactivated form of the kinase (Ki=0.04µM) compared to the activated form (Ki=4µM).


  • VEGFR inhibitor


Orange solid.


Soluble in DMSO (25mg/ml).

Formula: C₁₇H₁₆N₂O
Storage Temperature: Freezer
CAS Number: 5812-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbe8259f3c9b3ccc9e45f81021f1f86ba284eb06&vid=aebb7c224f6c7068a13a7d238e9760a91c2649b5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR