Tyrphostin AG-1296 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI303-0005 BML-EI303-0025
89158-010EA 267.02 CAD
89158-010 89158-012
Tyrphostin AG-1296 ≥98% (by TLC)
Tyrphostin AG-1296

A potent inhibitor of PDGF receptor tyrosine kinase (IC50=1 µM). Also inhibits FGF receptor tyrosine kinase and c-kit. Induces apoptosis in a small-cell lung cancer cell line (H526).


  • PDGF receptor tyrosine kinase inhibitor


Off-white to brownish solid.


Soluble in DMSO or dichloromethane.

Formula: C₁₆H₁₄N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00270913
CAS Number: 146535-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3351f057b02283a9b3f51c848f8893eb2920aa16&vid=df2b80a96ac69c2635452e45beee545760a55830&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR