5-Iodotubercidin ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 4-Amino-5-iodo-7-(β-D-ribofuranosyl)-pyrrolo[2,3-d]pyrimidine, (2R,3R,4S,5R)-2-(4-Amino-5-iodo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

BML-EI293-0001 BML-EI293-0005
89157-982EA 203.82 CAD
89157-982 89157-984
5-Iodotubercidin ≥98% (by TLC)
5-Iodotubercidin

Inhibits ERK2 (Ki = 525nM). Also inhibits adenosine kinase (Ki = 30nM), CK1 and CK2 and insulin receptor kinase (IC50 ranging from 0.4-28µM).


  • Protein kinase inhibitor


Off-white solid


Soluble in DMSO (25mg/ml) or 100% ethanol (8mg/ml warm)

Formula: C₁₁H₁₃IN₀₄
MW: 392.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00055131
CAS Number: 24386-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb27b21f0866c77e16549284b1b8ddecd8a38e6c&vid=f4272885a9418abbfee0a4e55cfeb0e2674011d7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR