U-0126 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI282-0001 BML-EI282-0005
89157-960EA 140.62 CAD
89157-960 89157-962
U-0126 ≥98% (by TLC)
U-0126

U-0126 (Bis[amino[(2-aminophenyl)thio]methylene]butanedinitrile) is a novel, potent and selective MEK inhibitor (MEK1; IC50=72nM, MEK2; IC50=58nM). Inhibits AP-1 transcription activity. Blocks the production of inflammatory cytokines and matrix metalloproteinases.


  • MEK inhibitor


White solid


Soluble in DMSO (>10mg/ml) or 100% ethanol (2mg/ml).

Formula: C₁₈H₁₆N₆S₂
Storage Temperature: Ambient
CAS Number: 109522-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e0351003991d1ba63f69398da99811c8740acca&vid=6e7c2e4b922ffd7450345358dc2fc89932193d19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR