BAY 11-7085 ≥99%

Supplier: Enzo Life Sciences

BML-EI279-0010 BML-EI279-0050
89157-952EA 183.28 CAD
89157-952 89157-954
BAY 11-7085 ≥99%
BAY 11-7085

NF-kappaB inhibitor


An irreversible inhibitor of cytokine-inducible IkappaBalpha phosphorylation (IC50=10 µM). In intact HUVECs it inhibits TNFalpha-induced phosphorylation and degradation of IkappaBalpha without affecting constitutive phosphorylation. Inhibition of IkappaBalpha phosphorylation and degradation results in the inactivation of NF-kappaB and NF-kappaB-dependent expression of endothelial-leukocyte adhesion molecules, E-selectin, VCAM-1 and ICAM-1. BAY 11-7085 displayed potent anti-inflammatory activity in the rat carrageenan paw edema assay and adjuvant arthritis model.

Formula: C₁₃H₁₅NO₂S
MDL Number: MFCD01862602
CAS Number: 196309-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff56d563ae3470e0b7e1767733a3ed0da4ed907c&vid=ed694ccdeb6570d3a7fc5a78ec8597929916c41f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR