BAY 11-7082 ≥99%

Supplier: Enzo Life Sciences

Synonyms: (E)-3-(p-Toluenesulphonyl)acrylonitrile, (E)-3-Tosylacrylonitrile, (E)-3-(p-Toluenesulfonyl)acrylonitrile

BML-EI278-0010 BML-EI278-0050
89157-948EA 183.28 CAD
89157-948 89157-950
BAY 11-7082 ≥99%
BAY 11-7082

NF-kappaB inhibitor


Inhibitor of cytokine induced IκBα phosphorylation resulting thereby in a decreased expression of NF-κB and adhesion molecules. Potential anti-inflammatory agent.

Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
MDL Number: MFCD00712162
CAS Number: 19542-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d5b16b04e310dcb3f47ff5bc7c6de19183b7155&vid=51d8e1d293ad0df26da25a9b8f5b5b36a01dd8fb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR