HBDDE ≥95% (by TLC)
Supplier: Enzo Life Sciences
BML-EI273-0001
BML-EI273-0005
89157-936EA
282.82
CAD
89157-936
89157-938
HBDDE ≥95% (by TLC)
HBDDE
Inhibits PKCα (IC50=43 µM) and PKCγ (IC50=50 µM) without inhibiting the δ, βI and βII isotypes.
- PKC inhibitor
Orange solid.
Soluble in DMSO (25 mg/ml), ethanol (15 mg/ml warm) or water (10 mg/ml warm).
Formula:
C₁₆H₁₈O₈ Melting Pt: 165…168 °C Storage Temperature: Freezer |
MDL Number:
MFCD00925616 CAS Number: 154675-18-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...