MK-886 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI266-0005 BML-EI266-0025
89157-920EA 176.96 CAD
89157-920 89157-922
MK-886 ≥98% (by HPLC)
MK-886

Inhibitor of PPARα activation. Inhibits 5-lipoxygenase-activating protein (FLAP) and leukotriene biosynthesis. Binds to FLAP with high affinity and prevents the activation of 5-lipoxygenase. IC50’s for leukotriene biosynthesis: 2.5 nM in intact leukocytes; 1.1 µM in whole blood. Induces apoptosis through the mitochondrial pathway.


  • FLAP inhibitor


White solid.


Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml).

Formula: C₂₇H₃₃ClNO₂S·Na
Storage Temperature: Ambient
MDL Number: MFCD00876710
CAS Number: 118414-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91a1291a05c96db156c21a172c0e177f09b0113f&vid=1bd1cbdece43c0726fdd53ac4546356678ec2c7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR