Tyrphostin AG-126 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-[(3-Hydroxy-4-nitrophenyl)methylidene]propanedinitrile, Tyrphostin AG 126, 2-(3-Hydroxy-4-nitrobenzylidene)malononitrile, AG-126, (3-Hydroxy-4-nitrobenzylidene)malononitrile

BML-EI263-0005 BML-EI263-0025
89157-912EA 183.28 CAD
89157-912 89157-914
Tyrphostin AG-126 ≥98% (by HPLC)
Tyrphostin AG-126

AG-126 prevents lipopolysaccharide-induced lethal toxicity (septic shock) in mice. Administered at 1.5 mg/mouse, it reduced LPS-induced lethal toxicity from 95% to 10%. AG-126 greatly reduces LPS-induced tyrosine phosphorylation of p42MAPK in murine peritoneal macrophages at 50 µM. EGFR, Her-2/neu receptor or PDGF-R are not inhibited at 100 µM. Inhibits tyrosine kinase-dependent B cell receptor signaling.


  • Inhibits tyrosine kinase-dependent B cell receptor signaling


Yellow solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (5mg/ml warm).

Formula: C₁₀H₅N₃O₃
Melting Pt: 181…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00236448
CAS Number: 118409-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6298223c475925f46712741e5195d64fe4a6d9ae&vid=1b6917c92d32148a7c38d78a80f41be6cb0900ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR