N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI261-0005 BML-EI261-0025
89157-904EA 188.02 CAD
89157-904 89157-906
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide ≥98% (by HPLC)
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulfonamide

Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity.


  • COX inhibitor


Cyclooxygenase inhibitor. Selectively inhibits PGH synthase-2 (IC50=1 µM) with no inhibition of PGH synthase-1 at concentrations up to 100 µM. Protects neuronal cultures from LPS-induced neurotoxicity.


Soluble in 100% ethanol (warm; sonication), DMSO (25mg/ml) or dimethyl formamide. Insoluble in water.

Formula: C₁₃H₁₈N₂O₅S
Storage Temperature: Ambient
MDL Number: MFCD00882995
CAS Number: 123653-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9213d5b3ded2e62b16d0d1929536f2b404d5630&vid=d2c7ad5aad8b3c6dc3d3d57bfb72e86684ee90e3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR