AG-879 ≥99%
Supplier: Enzo Life Sciences
Synonyms:
Tyrphostin AG 879, (E)-2-Cyano-3-(3,5-di-tert-butyl-4-hydroxyphenyl)prop-2-enethioamide
BML-EI258-0005
BML-EI258-0025
89157-892EA
181.7
CAD
89157-892
89157-894
AG-879 ≥99%
AG-879
NGF receptor inhibitor
AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells.
Formula:
C₁₈H₂₄N₂OS MW: 316.4610 g/mol |
MDL Number:
MFCD00236450 CAS Number: 148741-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...