Fasudil dihydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 1-(5-Isoquinolinesulfonyl)homopiperazine dihydrochloride salt, 1-(5-Isoquinolinesulphonyl)homopiperazine dihydrochloride salt, HA-1077 dihydrochloride salt

BML-EI233-0010 BML-EI233-0050
89157-834EA 267.02 CAD
89157-834 89157-836
Fasudil dihydrochloride ≥98% (by TLC)
Fasudil dihydrochloride

Inhibits Rho kinase (ROCK; Ki=0.33μM). Selective protein kinase A and G inhibitor (both Ki’s = 1.6μM). Inhibits agonist-induced activation of NADPH oxidase in human neutrophils (IC50=15μM). Inhibitor of myosin light chain kinase and Ca2+/calmodulin-dependent protein kinase II. Inhibits translocation of PKCβI, PKCβII and PKCζ. Cell permeable Ca2+ antagonist with antivasospastic properties.


  • Rho kinase (ROCK) inhibitor


White to off-white solid.


Soluble in water (>20mg/ml), methanol or DMSO.

Formula: C₁₄H₁₇N₃O₂S·2HCl
Melting Pt: 238…242 °C
Storage Temperature: Freezer
MDL Number: MFCD00153805
CAS Number: 203911-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=570d6af2bb518e1b47a898c3dc783743d80f0dfb&vid=d2fa5fec0e9d6080ed03f11d9ce8dec4660be0eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR