Tyrphostin B7 ≥99% (by TLC)
Supplier: Enzo Life Sciences
BML-EI229-0002
BML-EI229-0010
89157-820EA
227.52
CAD
89157-820
89157-822
Tyrphostin B7 ≥99% (by TLC)
Tyrphostin B7
Member of the tyrphostin family of tyrosine kinase inhibitors. Selective inhibitor of the PDGF receptor kinase (IC50=20µM) vs. the EGF receptor kinase (IC50=820µM). Inhibits PDGF-induced mitogenesis in human bone marrow fibroblasts.
- PDGF receptor inhibitor
Yellow solid.
Soluble in DMSO (25mg/ml) or 100% ethanol (2mg/ml, warm).
Formula:
C₁₅H₉N₅ Melting Pt: 300…302 °C Storage Temperature: Freezer |
MDL Number:
MFCD00236449 CAS Number: 134036-53-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...