Tyrphostin A9 ≥99% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: [4-Hydroxy-3,5-bis(2-methyl-2-propanyl)benzylidene]malononitrile, Malonoben

BML-EI215-0050 BML-EI215-0250
89157-792EA 146.94 CAD
89157-792 89157-794
Tyrphostin A9 ≥99% (by HPLC)
Tyrphostin A9

Tyrphostin 9 is a selective inhibitor of the PDGF receptor tyrosine kinase (IC50=1.2 µM). It is also a potent (10 nM) uncoupler of oxidative phosphorylation.


  • PDGF receptor inhibitor


Yellow solid.


Soluble in DMSO (>25mg/ml) or ethanol (20mg/ml).

Formula: C₁₈H₂₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00209853
CAS Number: 10537-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b650322c94c0ed0e241e857bb9af473ad1740fe&vid=d5153d7da389d78435ad30c86565eb697f4d917f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR