Cinnamyl-3,4-dihydroxy-α-cyanocinnamate ≥98%

Supplier: Enzo Life Sciences

BML-EI211-0010 BML-EI211-0050
89157-788EA 167.48 CAD
89157-788 89157-790
Cinnamyl-3,4-dihydroxy-α-cyanocinnamate ≥98%
Cinnamyl-3,4-dihydroxy-alpha-cyanocinnamate

Lipoxygenase inhibitor


Selective 12-lipoxygenase inhibitor. IC50=0.063 µM, 3.33 µM and 1.89 µM for 12-, 15- and 5-LO respectively in isolated enzyme preparations.

Formula: C₁₉H₁₅NO₄
MDL Number: MFCD00210839
CAS Number: 132465-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bce0f315c8510f238ac6354ae3a99b43fb10d64&vid=04d315b5449c5c467407fe88c0edfc42c0a307ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR