H-9 HCl ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI195-0010 BML-EI195-0050
89157-754EA 175.38 CAD
89157-754 89157-756
H-9 HCl ≥98% (by TLC)
H-9 HCl

An excellent affinity ligand for chromatographic purification of protein kinases. Inhibits protein kinases (Ki's: PKA=1.9 µM; PKC=18 µM; PKG=0.87 µM; MLCK=70 µM).


  • PKA inhibitor


White solid.


Soluble in water.

Formula: C₁₁H₁₃N₃O₂S·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00083242
CAS Number: 116970-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=209d3692b2f5e39db3cc554264e15ba1f2ec8673&vid=5144f450e435a399c768907f7544f0656c7d8813&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR