Tyrphostin 1 ≥99%

Supplier: Enzo Life Sciences

BML-EI190-0020 BML-EI190-0100
89157-742EA 129.56 CAD
89157-742 89157-744
Tyrphostin 1 ≥99%
Tyrphostin 1

An inactive tyrphostin, which may be used as a negative control.


  • Formulation: Yellow solid.


Soluble in 100% ethanol (25 mg/mL, warm) or DMSO (25 mg/mL).

Formula: C₁₁H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD00019787
CAS Number: 2826-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1db17f422db415ed0204addb7a770a1ceb99dac2&vid=ef3572333104d4cedb53d929035193baf3efd8a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR