H-7 dihydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 5-((2-Methylpiperazin-1-yl)sulphonyl)isoquinoline dihydrochloride, 5-((2-Methylpiperazin-1-yl)sulfonyl)isoquinoline dihydrochloride, Protein kinase inhibitor H-7 2HCl

BML-EI148-0010 BML-EI148-0050
89157-654EA 181.7 CAD
89157-654 89157-656
H-7 dihydrochloride ≥98% (by TLC)
H-7 dihydrochloride

Inhibits PKA (Ki=3.0μM), PKG (Ki=5.8μM), MLCK (Ki=97μM) and PKC (Ki=6μM). Also inhibits myosin light chain kinase. Induces apoptotic DNA fragmentation and cell death. Inhibits telomerase activity in treated NPC-076 cells. Cell permeable.


  • PKC inhibitor


White to light yellow solid.


Soluble in water (50mg/ml), 100% ethanol or DMSO.

Formula: C₁₄H₁₇N₃O₂S·2HCl
MW: 364.29 g/mol
Melting Pt: 219…222 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036961
CAS Number: 108930-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5f2af9ad323b668b4907d910e9e6bc88d3e4852&vid=54ddb51a517f236c3ff609281177431a76bc8d8b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR