RO 20-1724 ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-EI117-0100 BML-EI117-1000
89157-596EA 248.06 CAD
89157-596 89157-598
RO 20-1724 ≥99% (by TLC)
RO 20-1724

Selective, potent cAMP-specific phosphodiesterase inhibitor (PDE IV, IC50=2µM). Inhibits superoxide generation and platelet aggregation stimulated by arachidonic acid. Also inhibits fMLP-induced neutrophil adhesion to vascular endothelial cells.


  • PDE inhibitor


White to off-white solid.


Soluble in 100% ethanol (40mg/ml) or DMSO (200 mg/ml); insoluble in water.

Formula: C₁₅H₂₂N₂O₃
Melting Pt: 126…127 °C
Storage Temperature: Ambient
CAS Number: 29925-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eda329397442e9c5b37101d7fea8a78a8efc1972&vid=80057058e4443d4615d3f7032db610fbe38c3fd0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR