Ketoconazole ≥99%
Supplier: Enzo Life Sciences
Synonyms:
(+/-)-cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, cis-1-Acetyl-4-[4-[[2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine
Cytochrome P450 inhibitor
Inhibits cytochrome P-450-dependent steps in the biosynthesis of steroid hormones in vivo. Antimetastatic and antineoplastic activity. Antipsoriatic activity. Orally active 5-lipoxygenase and thromboxane synthase inhibitor.
Formula:
C₂₆H₂₈Cl₂N₄O₄ MW: 531.43 g/mol |
MDL Number:
MFCD00058579 CAS Number: 65277-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...