kainic acid ≥98% (by HPLC), white to off-white solid
Supplier: Enzo Life Sciences
Synonyms:
(2S,3S,4S)-3-(Carboxymethyl)-4-(prop-1-en-2-yl)pyrrolidine-2-carboxylic acid
BML-EA123-0005
BML-EA123-0025
89157-458EA
145.36
CAD
89157-458
89157-460
kainic acid ≥98% (by HPLC), white to off-white solid
kainic acid
Selective agonist at kainate receptors.
- Formulation: White to off-white solid.
Soluble in water (25 mg/mL).
Formula:
C₁₀H₁₅NO₄ MW: 213.233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077806 CAS Number: 487-79-6 |
Specification Test Results
Purity | ≥98% (HPLC) |
Identity | Determined by H-NMR |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...