25-Hydroxyvitamin D3 ≥98%, white to off-white solid

Supplier: Enzo Life Sciences
Danger

Synonyms: Calcifediol, (R,Z)-3-(2-((1R,3aS,7aR,E)-1-((R)-6-hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexan-1-ol

BML-DM100-0001 BML-DM100-0005
89157-372EA 140.62 CAD
89157-372 89157-374
25-Hydroxyvitamin D3 ≥98%, white to off-white solid
25-Hydroxyvitamin D3

Precursor to 1α,25-dihydroxyvitamin D₃


  • Formulation: White to off-white crystalline solid.


Intermediate in the biosynthesis of 1alpha,25-dihydroxyvitamin D3. Major circulating metabolite of vitamin D3.


Soluble in 100% ethanol.

Formula: C₂₇H₄₄O₂
MW: 400.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00867077
CAS Number: 19356-17-3
UN: 2811
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3ec23887b3d8e238fd5ef1c6f5940df673eb623&vid=ce4a390b2c87e0097d88cb0406310e7e5e60a035&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Determined by IR
Purity (HPLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR