(±)1,2-Didecanoylglycerol (10:0) ≥98% (by TLC), white solid

Supplier: Enzo Life Sciences

BML-DG100-0010 BML-DG100-0050
89157-342EA 148.52 CAD
89157-342 89157-344
(±)1,2-Didecanoylglycerol (10:0) ≥98% (by TLC), white solid
(±)1,2-Didecanoylglycerol (10:0)

PKC activator.


  • Formulation: White solid.


A membrane permeable but non-physiologic DAG. Stimulates protein kinase C in intact platelets.

Formula: C₂₃H₄₄O₅
Storage Temperature: Freezer
CAS Number: 17863-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f4dd48353838e67f89a3e47c1b52bcb2ced135e&vid=4dbe2501938959d51e1ee1cb57099e1d7301a280&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR