Anti-Dishevelled-2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
DA4270-0025 DA4270-0100
89157-330EA 222.78 CAD
89157-330 89157-332
Anti-Dishevelled-2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse dishevelled-2.

Type: Primary
Antigen: DISHEVELLED-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR