Clothiapine ≥99%

Supplier: Enzo Life Sciences
Danger

BML-D108-0020 BML-D108-0100
89157-296EA 67.94 CAD
89157-296 89157-298
Clothiapine ≥99%
Clothiapine

Dopamine receptor ligand.


  • Formulation: Off-white solid (m.p. 118-120°C).


A typical neuroleptic agent related to clozapine.


Soluble in DMSO (10 mg/mL).

Formula: C₁₈H₁₈ClN₃S
CAS Number: 2058-52-8
EINECS: 218-162-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0ff80f6be71604ba45bc998cf2c4067ea31279c&vid=549d6bad0e8cf82b31db973b78ea8a99c34fee49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR