Thiolutin (from Streptomyces luteosporeus) ≥95% (by HPLC, TLC)

Supplier: Enzo Life Sciences

BML-CT107-0001 BML-CT107-0005
89157-260EA 237 CAD
89157-260 89157-262
Thiolutin (from Streptomyces luteosporeus) ≥95% (by HPLC, TLC)
Thiolutin

Sulfur containing antibiotic. Potent inhibitor of bacterial and yeast RNA polymerases. Inhibits mRNA chain elongation. Inhibits adhesion of endothelial cells (HUVECs) to vitronectin (IC50=0.83 µM) thereby inhibiting tumor cell-induced angiogenesis in vivo. Inhibits transcription in C. albicans. Inhibits microbiological growth in beer. Active against soil borne pathogens.


  • RNA polymerase inhibitor


Yellow crystalline solid.


Soluble in DMSO (5mg/ml), very slightly soluble in water

Formula: C₈H₈N₂O₂S₂
Storage Temperature: Freezer
CAS Number: 87-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2298e1ff9f7adcca03e46cf7a6e71f77993503a4&vid=9f8071be697d512a5c720bf6ab2770eb28e9f3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR