WIN-55,212-2 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-CR105-0010 BML-CR105-0050
89157-106EA 178.54 CAD
89157-106 89157-108
WIN-55,212-2 ≥98% (by HPLC)
WIN-55,212-2

Cannabinoid receptor agonist.


  • Formulation: White to off-white solid.


High affinity cannabinoid receptor (CB) agonist (Ki = 62.3 nM and 3.3 nM at the human cloned CB₁ and CB₂ receptors, respectively). Potent analgesic in a rat model of neuropathic pain. Activates p42 and p44 MAP kinase via a receptor-mediated signaling.


Soluble in DMSO, insoluble in water.


Synonym: (R)-(+)-[2,3-Dihydro-5-methyl-3-(4-morpholinylmethyl)pyrrolo[1,2,3-de]-1,4-benzoxazin-6-yl]-1-naphthalenylmethanone.Mesylate.

Formula: C₂₇H₂₆N₂O₃·CH₃SO₃H
Storage Temperature: Refrigerator
CAS Number: 131543-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffbeab5329ec79c58e7368bcb5b2971e17f321a1&vid=1e86711934d4624e12e33c86a9261d24eeea9d36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR