CP-55,940 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-CR102-0005 BML-CR102-0025
89157-098EA 353.92 CAD
89157-098 89157-100
CP-55,940 ≥98% (by HPLC)
CP-55,940

Cannabinoid receptor ligand.


  • Formulation: White solid.


Cannabinoid CB₁ and CB₂ agonist which is more potent than δ9-THC. Kd’s for cloned/transfected CB₁ and CB₂: 2.6 and 3.7 nM. Has been reported to induce the immediate early gene Krox-24 in corpus striatum and cultured astrocytes. Stimulates proliferation of stimulated B-cells at low nM concentrations.


Soluble in DMSO (4 mg/mL) or ethanol.

Formula: C₂₄H₄₀O₃
Storage Temperature: Freezer
CAS Number: 83002-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85c32058a45768dae873b7fe96a6ad7bf494ccc3&vid=a1976f92060d14c8465a562780c7be737cd98b43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR and MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR