8-Br-cGMP ≥96% (by HPLC)

Supplier: Enzo Life Sciences

BML-CN205-0010 BML-CN205-0050
89157-028EA 140.62 CAD
89157-028 89157-030
8-Br-cGMP ≥96% (by HPLC)
8-Br-cGMP

A cell-permeable cGMP analog which is more resistant to phosphodiesterases than cGMP and which preferentially activates cGMP-dependent protein kinase. It inhibits thrombin stimulated arachidonic acid release in human platelets.


  • PKG activator


White to off-white powder


Soluble in water (25mg/ml) or aqueous buffers. Sparingly soluble in methanol (4mM). Insoluble in DMSO, dimethyl formamide and 96% ethanol.

Formula: C₁₀H₁₂BrN₅NaO₈P
Storage Temperature: Freezer
CAS Number: 51116-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ae97b73b1204d05a6117abe87242bcd80ec00d8&vid=3f6827122ac1cb58dff9e2e731bec69ef589ce9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR