8-Bromo-cAMP sodium salt ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 8- Bromoadenosine- 3', 5'- cyclic monophosphate, 8-Br-cAMP, 8-Br-Camp sodium salt, 8-Bromoadenosine 3',5'-cyclic monophosphate sodium salt, 8-Bromo-cAMP, 8- Bromoadenosine- 3', 5'-monophosphate

BML-CN115-0010 BML-CN115-0050
89157-000EA 113.76 CAD
89157-000 89157-002
8-Bromo-cAMP sodium salt ≥98% (by HPLC)
8-Bromo-cAMP sodium salt

PKA activator.


  • Formulation: White to grey powder


A cell-permeable cAMP analog which is more resistant to phosphodiesterases than cAMP and which preferentially activates cAMP-dependent protein kinase (PKA). Has increased metabolic stability and membrane permeability comapred with cAMP.


Soluble in water (100 mg/mL).

Formula: C₁₀H₁₀BrN₅NaO₆P
Storage Temperature: Freezer
MDL Number: MFCD00005844
CAS Number: 76939-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce7051753c7d250d077cb5682d455c0a19e124da&vid=5c801f0106550354358301d392a728a41c3815e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR, ³¹P-NMR, ESI-MS and UV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR