2',5'-Dideoxyadenosine ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-CN110-0005 BML-CN110-0025
89156-996EA 237 CAD
89156-996 89156-998
2',5'-Dideoxyadenosine ≥98% (by TLC)
2',5'-Dideoxyadenosine

Adenylate cyclase inhibitor


A useful, cell-permeable inhibitor of adenylate cyclase (ID50=250µM). Inhibits growth and caues cell differentiation (1.5µM) in K562 cells.

Formula: C₁₀H₁₃N₅O₂
MDL Number: MFCD00210904
CAS Number: 6698-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ab68638f143149a79ebd3bac821ea05e3418c80&vid=cc06f876d932bc8df02eac72521b6066164f9ad3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR