Embelin ≥98%

Supplier: Enzo Life Sciences

BML-CM125-0010 BML-CM125-0050
89156-972EA 150.1 CAD
89156-972 89156-974
Embelin ≥98%
Embelin

XIAP inhibitor


Embelin is a novel cell-permeable inhibitor of XIAP (X-linked inhibitor of apoptosis) which was discovered by screening a library of natural products derived from traditional Chinese medicine. It binds to the XIAP BIR3 domain preventing its binding to and inhibition of caspase-9 and effector caspases. Embelin selectively inhibits cell growth in cancer cells expressing high levels of XIAP (IC50 = 3-6 µM) versus normal cells with low levels of XIAP.

Formula: C₁₇H₂₆O₄
MDL Number: MFCD00016369
CAS Number: 550-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e842c3874128f7a4960387bba359a264d38ef6&vid=0e4a11c7f9101171427beb99759ad72712f95639&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR