2-Methoxy-antimycin A ≥85%
Supplier: Enzo Life Sciences
BML-CM123-0001 BML-CM123-0005
89156-966EA 146.94 CAD
89156-966 89156-968
2-Methoxy-antimycin A ≥85%
2-Methoxy-antimycin A
An analog of antimycin A (mixture of antimycin A1, A2, A3 and A4) which binds to the hydrophobic groove of Bcl-2 and Bcl-XL thereby inducing apoptosis
It displays selective cytotoxicity for Bcl-XL-expressing cells (10 µg/ml). Unlike antimycin A it does not exhibit general mitochondrial toxicity.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c4aeb38e6582c61d0895ba6bdf4d27b62bdfddb&vid=67106b363bd1e39b89ac1e7c1de88eaffa6c4ded&prid=web&clid=SAPDAM