FCCP ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
FCCP, [4-(Trifluoromethoxy)phenyl]carbonohydrazonoyl dicyanide, [4-(Trifluoromethoxy)phenylhydrazono]malononitrile, Carbonyl cyanide 4-(Trifluoromethoxy)phenylhydrazone
MPTP inhibitor
A potent reversible inhibitor of mitochondrial oxidative phosphorylation. FCCP is a useful tool for depolarizing mitochondrial membrane potential. Treatment of cells with FCCP at varying concentrations leads to partial (100 nM) or complete (10 µM) depolarization and apoptosis.
Formula:
C₁₀H₅F₃N₄O MW: 254.17 g/mol |
MDL Number:
MFCD00009699 CAS Number: 370-86-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...