CGP-37157 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: CGP37157, 7-Chloro-5-(2-chlorophenyl)-1,5-dihydrobenzo[e][1,4]thiazepin-2(3H)-one

BML-CM119-0005 BML-CM119-0025
89156-958EA 219.62 CAD
89156-958 89156-960
CGP-37157 ≥98% (by TLC)
CGP-37157

Potent, selective inhibitor of mitochondrial Na+Ca2+ exchange. IC50=0.4µM. Blocks mitochondrial permeability transition.


  • MPTP inhibitor


White solid.


Soluble in DMSO.

Formula: C₁₅H₁₁Cl₂NOS
MW: 324.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09055422
CAS Number: 75450-34-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c687b20337f441b6de3683db0edcac8c57747d8&vid=687a285689745c5ea3de1e1616d7df68e288b388&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR