Citrinin ≥98%

Supplier: Enzo Life Sciences

Synonyms: Citrinin, yellow, (3S,4R)-8-hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid

BML-CM116-0005 BML-CM116-0025
89156-950EA 200.66 CAD
89156-950 89156-952
Citrinin ≥98%
Citrinin

MPTP activator


Antibiotic. Induces mitochondrial permeability pore opening and inhibits respiration by interfering with complex I of the respiratory chain. Mycotoxin. Acts as nephrotoxin in all species in which it has been tested. Has been implicated as a cause of Balkan nephropathy and yellow rice fever in humans. Induces apoptosis.


Isolated from Penicillium citrinum

Formula: C₁₃H₁₄O₅
MW: 250.25 g/mol
MDL Number: MFCD00006912
CAS Number: 518-75-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0003a66f4db2b0401a2ba8ff27ff5d847cd9a44&vid=2a351bcd7ffbb539ea5f9a526d230c345a556788&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR