Decylubiquinone ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-CM115-0010 BML-CM115-0050
89156-946EA 131.14 CAD
89156-946 89156-948
Decylubiquinone ≥98% (by TLC)
Decylubiquinone
MPTP inhibitor
Inhibits the Ca2+ dependent mitochondrial permeability transition pore (MPTP) without inhibiting respiration. Maximum effect on Ca2+ retention capacity of rat liver mitochondria at 100μM. Inhibits etoposide-induced apoptosis in in L929 cells at 5μM.
CAS Number: 55486-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=766a9476c7a8d3f22efafa43ff50fe68eac75b2d&vid=086ba947302ecac492332d221e3d041dc09127e0&prid=web&clid=SAPDAM