Bongkrekic acid triammonium salt ≥92% (by HPLC)

Supplier: Enzo Life Sciences

BML-CM113-0100 BML-CM113-1000
89156-940EA 300.2 CAD
89156-940 89156-942
Bongkrekic acid triammonium salt ≥92% (by HPLC)
Bongkrekic acid triammonium salt

Bongkrekic acid is an inhibitory ligand of the mitochondrial adenine nucleotide translocase. It inhibits mitochondrial permeability transition pore opening. At 50 µM it inhibits dexamethasone- apoptosis in thymocytes, preventing the PARP cleavage and DEVDase (caspase-3 like) activity normally produced by these treatments.


  • MPTP inhibitor


Beige to pink lyophilized solid.

Storage Temperature: Freezer
CAS Number: 11076-19-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=339af1e032e8e25e1f8b64557cc9ec23f9ca630f&vid=7c2b3f8b423f4919bfa1aa909ff3f5a9457e0318&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR