[6]-Gingerol ≥97% (by HPLC)

Supplier: Enzo Life Sciences
Danger

BML-CA422-0005 BML-CA422-0020
89156-872EA 244.9 CAD
89156-872 89156-874
[6]-Gingerol ≥97% (by HPLC)
[6]-Gingerol

ATPase activator.


  • Formulation: White to pale yellow oily or waxy solid.


Gingerol ([6]-gingerol), a pungent vanilloid isolated from ginger rhizomes (Zingiber officinale), is a novel and selective activator of skeletal and cardiac SR Ca²⁺-ATPase (EC₅₀ = 4 µM). It is a novel cardiotonic agent with antiplatelet activity. It induces apoptosis in HL-60 cells. Acts as an agonist at vanilloid receptors.


Soluble in 100% ethanol.

Formula: C₁₇H₂₆O₄
Storage Temperature: Freezer
MDL Number: MFCD00210507
CAS Number: 23513-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38dfa9b657449eb7707598842e9f879d5bb55477&vid=2216de997a3bb0e6f2c6cd8867df575b9168951e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR